(Benzoylmethylene)triphenylphosphorane ≥98.0%

Supplier: TCI America

Synonyms: 1-Phenyl-2-(triphenylphosphoranylidene)ethanone

T2002-5G T2002-1G
TCT2002-5GEA 323.9 CAD
TCT2002-5G 76220-516
(Benzoylmethylene)triphenylphosphorane ≥98.0%
(Benzoylmethylene)triphenylphosphorane

CAS Number: 859-65-4
MDL Number: MFCD00014088
Molecular Formula: C26H21OP
Molecular Weight: 380.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 188
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Benzoylmethylene)triphenylphosphorane

Formula: C₂₆H₂₁OP
MW: 380.43 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00014088
CAS Number: 859-65-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53cbffdc7169a2bd9905f19acb05df5937646eed&vid=53cbffdc7169a2bd9905f19acb05df5937646eed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR