(Triphenylphosphoranylidene)acetaldehyde ≥98.0% (by HPLC)

Supplier: TCI America

T2001-5G T2001-25G
TCT2001-5GEA 123.24 CAD
TCT2001-5G TCT2001-25G
(Triphenylphosphoranylidene)acetaldehyde ≥98.0% (by HPLC)
(Triphenylphosphoranylidene)acetaldehyde

CAS Number: 2136-75-6
MDL Number: MFCD00006994
Molecular Formula: C20H17OP
Molecular Weight: 304.33
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 198
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Formylmethylene)triphenylphosphorane

Formula: C₂₀H₁₇OP
Melting Pt: 198 °C
MDL Number: MFCD00006994
CAS Number: 2136-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e48f37235609ce09dc0aebab7fadc0214183952&vid=6e48f37235609ce09dc0aebab7fadc0214183952&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR