(±)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0%

Supplier: TCI America

Synonyms: Tetralin-1-amine, 1,2,3,4-Tetrahydronaphthalen-1-amine

T1992-5G T1992-25G
TCT1992-5GEA 185.18 CAD
TCT1992-5G TCT1992-25G
(±)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0%
(±)-1,2,3,4-Tetrahydro-1-naphthylamine

CAS Number: 2217-40-5
MDL Number: MFCD00001740
Molecular Formula: C10H13N
Molecular Weight: 147.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 230
Flash Point (°C): 113
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 1-Amino-1,2,3,4-tetrahydronaphthalene

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 230 °C
Density: 1.04 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00001740
CAS Number: 2217-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a58139be919b2c5e3266a8565ad7508378e11003&vid=a58139be919b2c5e3266a8565ad7508378e11003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR