3-Thiopheneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3-Thienylboronic acid, Thiophene-3-boronic acid
T1975-1G
T1975-5G
TCT1975-1GEA
44.24
CAD
TCT1975-1G
TCT1975-5G
3-Thiopheneboronic acid (contains varying amounts of Anhydride)
3-Thiopheneboronic acid
CAS Number: 6165-69-1
MDL Number: MFCD00151851
Molecular Formula: C4H5BO2S
Molecular Weight: 127.95
Form: Crystal
Color: White
Other Product Names / Synonyms: 3-Thienylboronic Acid
Formula:
C₄H₅BO₂S MW: 127.96 g/mol |
MDL Number:
MFCD00151851 CAS Number: 6165-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...