4'-Methyl-2,2,2-trifluoroacetophenone ≥96.0%
Supplier: TCI America
Synonyms:
2,2,2-Trifluoro-1-(p-tolyl)ethanone, 4-(Trifluoroacetyl)toluene, 2,2,2-Trifluoro-1-(4-methylphenyl)ethan-1-one
CAS Number: 394-59-2
MDL Number: MFCD00052338
Molecular Formula: C9H7F3O
Molecular Weight: 188.15
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.23
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4'-Methyl-2,2,2-trifluoroacetophenone, 4-(Trifluoroacetyl)toluene
Formula:
C₉H₇F₃O MW: 188.15 Density: 1.23 (20 °C) |
MDL Number:
MFCD00052338 CAS Number: 394-59-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...