2,4,6-Trimethylphenylboronic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

T1960-1G T1960-5G
TCT1960-1GEA 46.8 CAD
TCT1960-1G TCT1960-5G
2,4,6-Trimethylphenylboronic acid ≥98.0% (by HPLC)
2,4,6-Trimethylphenylboronic acid

CAS Number: 5980-97-2
MDL Number: MFCD00236060
Molecular Formula: C9H13BO2
Molecular Weight: 164.01
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-Mesityleneboronic Acid, 2,4,6-Trimethylbenzeneboronic Acid

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
MDL Number: MFCD00236060
CAS Number: 5980-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=521e665a6d2dd56c7b1eb83598ec91dbc50aa0b7&vid=521e665a6d2dd56c7b1eb83598ec91dbc50aa0b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR