3-(Trifluoromethoxy)benzyl alcohol ≥98.0%
Supplier: TCI America
T1955-5G
T1955-1G
TCT1955-5GEA
276.03
CAD
TCT1955-5G
TCT1955-1G
3-(Trifluoromethoxy)benzyl alcohol ≥98.0%
3-(Trifluoromethoxy)benzyl alcohol
CAS Number: 50823-90-0
MDL Number: MFCD00061270
Molecular Formula: C8H7F3O2
Molecular Weight: 192.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.32
Formula:
C₈H₇F₃O₂ MW: 192.14 g/mol Density: 1.32 (20 °C) |
MDL Number:
MFCD00061270 CAS Number: 50823-90-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...