4-Hydroxyphenylboronic acid pinacol ester ≥97.0%
Supplier: TCI America
T1954-5G
T1954-1G
TCT1954-5GEA
234.79
CAD
TCT1954-5G
TCT1954-1G
4-Hydroxyphenylboronic acid pinacol ester ≥97.0%
4-Hydroxyphenylboronic acid pinacol ester
CAS Number: 269409-70-3
MDL Number: MFCD02093756
Molecular Formula: C12H17BO3
Molecular Weight: 220.08
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 118
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Hydroxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Hydroxyphenylboronic Acid Pinacol Ester
Formula:
C₁₂H₁₇BO₃ Melting Pt: 118 °C |
MDL Number:
MFCD02093756 CAS Number: 269409-70-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...