4-Aminophenylboronic acid pinacol ester ≥98.0%

Supplier: TCI America

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

T1951-5G T1951-1G
TCT1951-5GEA 271.76 CAD
TCT1951-5G TCT1951-1G
4-Aminophenylboronic acid pinacol ester ≥98.0%
4-Aminophenylboronic acid pinacol ester

CAS Number: 214360-73-3
MDL Number: MFCD02093721
Molecular Formula: C12H18BNO2
Molecular Weight: 219.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 170


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Aminophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Aminophenylboronic Acid Pinacol Ester

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Melting Pt: 170 °C
MDL Number: MFCD02093721
CAS Number: 214360-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a870f77a0cfff92844b16219ee5475dc5bbcb04&vid=6a870f77a0cfff92844b16219ee5475dc5bbcb04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR