Ethyl-2-(triphenylphosphoranylidene)acetate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate
CAS Number: 1099-45-2
MDL Number: MFCD00009183
Molecular Formula: C22H21O2P
Molecular Weight: 348.38
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 130
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Carbethoxymethylene)triphenylphosphorane, (Ethoxycarbonylmethylene)triphenylphosphorane, (Triphenylphosphoranylidene)acetic Acid Ethyl Ester
Formula:
C₂₂H₂₁O₂P MW: 348.38 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00009183 CAS Number: 1099-45-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...