2,4,5-Trifluoro-3-methoxybenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: Moxidectin Impurity 5, 3-Methoxy-2,4,5-trifluorobenzoic acid

T1917-25G T1917-5G
TCT1917-25GEA 39.5 CAD
TCT1917-25G TCT1917-5G
2,4,5-Trifluoro-3-methoxybenzoic acid ≥98.0%
2,4,5-Trifluoro-3-methoxybenzoic acid

CAS Number: 112811-65-1
MDL Number: MFCD00153201
Molecular Formula: C8H5F3O3
Molecular Weight: 206.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 118


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4,5-Trifluoro-m-anisic Acid

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00153201
CAS Number: 112811-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fd0fcef9ff0b2943b0e920249ca97bb3fea78e1&vid=8fd0fcef9ff0b2943b0e920249ca97bb3fea78e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR