(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
(3R,4S,5R,6R)-3,4,5-tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-glucopyranose
T1914-1G
T1914-5G
TCT1914-1GEA
63.2
CAD
TCT1914-1G
TCT1914-5G
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol ≥95.0% (by HPLC)
(3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol
CAS Number: 4132-28-9
MDL Number: MFCD00066004
Molecular Formula: C34H36O6
Molecular Weight: 540.66
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Formula:
C₃₄H₃₆O₆ MW: 540.65 g/mol |
MDL Number:
MFCD00066004 CAS Number: 4132-28-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...