2',4',6'-Trihydroxyacetophenone monohydrate ≥98.0%

Supplier: TCI America

Synonyms: 2-Acetylphloroglucinol monohydrate, THAP monohydrate, Phloroacetophenone monohydrate

T1888-5G T1888-25G
TCT1888-5GEA 126.4 CAD
TCT1888-5G TCT1888-25G
2',4',6'-Trihydroxyacetophenone monohydrate ≥98.0%
2',4',6'-Trihydroxyacetophenone monohydrate

CAS Number: 480-66-0
MDL Number: MFCD00002287
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 215


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2-Acetylphloroglucinol, Phloroacetophenone

Formula: C₈H₈O₄·1H₂O
Melting Pt: 215 °C
MDL Number: MFCD00149091
CAS Number: 207-556-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b203f2ecee303aabd753f83adce87e85fdf6e1e&vid=1b203f2ecee303aabd753f83adce87e85fdf6e1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR