2,3,5,6-Tetrafluorobenzyl alcohol ≥96.0%

Supplier: TCI America

T1884-1G T1884-5G
TCT1884-1GEA 50.56 CAD
TCT1884-1G TCT1884-5G
2,3,5,6-Tetrafluorobenzyl alcohol ≥96.0%
2,3,5,6-Tetrafluorobenzyl alcohol

CAS Number: 4084-38-2
MDL Number: MFCD00792428
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 33

Formula: C₇H₄F₄O
MDL Number: MFCD00792428
CAS Number: 4084-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87403080fcefe9e6849b279fdbdc45b000abd9f1&vid=87403080fcefe9e6849b279fdbdc45b000abd9f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR