2,3,4,5-Tetrafluorobenzyl alcohol ≥98.0%

Supplier: TCI America

T1883-1G T1883-5G
TCT1883-1GEA 71.1 CAD
TCT1883-1G TCT1883-5G
2,3,4,5-Tetrafluorobenzyl alcohol ≥98.0%
2,3,4,5-Tetrafluorobenzyl alcohol

CAS Number: 53072-18-7
MDL Number: MFCD01631452
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 101

Formula: C₇H₄F₄O
Flash Pt: 101 °C
MDL Number: MFCD01631452
CAS Number: 53072-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=966e508266c1d651a44cf71ec83c9bdfb9caa417&vid=966e508266c1d651a44cf71ec83c9bdfb9caa417&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR