2,3,4-Trifluorobenzoic acid
Supplier: TCI America
T1867-1G
T1867-5G
T1867-25G
TCT1867-1GEA
26.86
CAD
TCT1867-1G
TCT1867-5G
TCT1867-25G
2,3,4-Trifluorobenzoic acid
2,3,4-Trifluorobenzoic acid
CAS Number: 61079-72-9
MDL Number: MFCD00061232
Molecular Formula: C7H3F3O2
Molecular Weight: 176.09
Form: Crystal
Color: White
Melting point (°C): 141
Formula:
C₇H₃F₃O₂ Melting Pt: 141 °C |
MDL Number:
MFCD00061232 CAS Number: 61079-72-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...