3-(Trifluoromethoxy)cinnamic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-(3-(Trifluoromethoxy)phenyl)acrylic acid

T1780-25G T1780-5G
TCT1780-25GEA 737.23 CAD
TCT1780-25G TCT1780-005G
3-(Trifluoromethoxy)cinnamic acid ≥98.0%
3-(Trifluoromethoxy)cinnamic acid

CAS Number: 168833-80-5
MDL Number: MFCD00066337
Molecular Formula: C10H7F3O3
Molecular Weight: 232.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 95

Formula: C₁₀H₇F₃O₃
MW: 232.16 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00066337
CAS Number: 168833-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed9be395fd3315231f3d640344c651217b452a6a&vid=ed9be395fd3315231f3d640344c651217b452a6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR