1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥95.0%

Supplier: TCI America

Synonyms: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

T1770-25G T1770-5G
TCT1770-25GEA 533.88 CAD
TCT1770-25G TCT1770-005G
1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥95.0%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane

CAS Number: 51851-37-7
MDL Number: MFCD00042333
Molecular Formula: C14H19F13O3Si
Molecular Weight: 510.37
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 86
Flash Point (°C): 84
Specific Gravity (20/20): 1.34


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 1H,1H,2H,2H-Tridecafluoro-n-octyltriethoxysilane, Triethoxy-1H,1H,2H,2H-perfluoro-n-octylsilane

Formula: C₁₄H₁₉F₁₃O₃Si
Boiling Pt: 86 °C
Density: 1.34 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00042333
CAS Number: 51851-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aff85c715a0eb10abcc887b0ec0ff7dc7d2781f&vid=3aff85c715a0eb10abcc887b0ec0ff7dc7d2781f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR