4-(p-Tolyl)butyric acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

T1727-5G T1727-1G T1727-25G
TCT1727-5GEA 147.1 CAD
TCT1727-5G TCT1727-001G TCT1727-25G
4-(p-Tolyl)butyric acid ≥96.0% (by GC, titration analysis)
4-(p-Tolyl)butyric acid

CAS Number: 4521-22-6
MDL Number: MFCD00021786
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 150
Melting point (°C): 58

Formula: C₁₁H₁₄O₂
Boiling Pt: 150 °C
Melting Pt: 58 °C
MDL Number: MFCD00021786
CAS Number: 4521-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=602a861a550d0ea865b34d9f1171fa5da3de351e&vid=602a861a550d0ea865b34d9f1171fa5da3de351e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR