1,1,2-Trimethyl-1H-benz[e]indole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1,1,2-Trimethyl-1H-benzo[e]indole
T1714-5G
T1714-25G
T1714-100G
TCT1714-005GEA
90.06
CAD
TCT1714-005G
TCT1714-025G
76220-504
1,1,2-Trimethyl-1H-benz[e]indole ≥98.0% (by HPLC, titration analysis)
1,1,2-Trimethyl-1H-benz[e]indole
CAS Number: 41532-84-7
MDL Number: MFCD00082627
Molecular Formula: C15H15N
Molecular Weight: 209.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 115
Other Product Names / Synonyms: 2,3,3-Trimethyl-4,5-benzo-3H-indole, 1,1,2-Trimethylnaphtho[1,2-d]pyrrole
Formula:
C₁₅H₁₅N MW: 209.29 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00082627 CAS Number: 41532-84-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...