3-(Trifluoromethyl)phenylacetone ≥95.0%

Supplier: TCI America

Synonyms: 1-[3-(Trifluoromethyl)phenyl]-2-propanone

T1693-25G T1693-5G
TCT1693-025GEA 110.6 CAD
TCT1693-025G TCT1693-5G
3-(Trifluoromethyl)phenylacetone ≥95.0%
3-(Trifluoromethyl)phenylacetone

CAS Number: 21906-39-8
MDL Number: MFCD00000397
Molecular Formula: C10H9F3O
Molecular Weight: 202.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 90
Specific Gravity (20/20): 1.22

Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
Density: 1.22 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00000397
CAS Number: 21906-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f394aa1e7a583184a8f82993550513c5c3d9b77&vid=6f394aa1e7a583184a8f82993550513c5c3d9b77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR