1-(Triisopropylsilyl)pyrrole ≥96.0%

Supplier: TCI America

T1677-5ML T1677-25ML
TCT1677-005MLEA 167.48 CAD
TCT1677-005ML TCT1677-025ML
1-(Triisopropylsilyl)pyrrole ≥96.0%
1-(Triisopropylsilyl)pyrrole

CAS Number: 87630-35-1
MDL Number: MFCD00054932
Molecular Formula: C13H25NSi
Molecular Weight: 223.44
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 107
Specific Gravity (20/20): 0.91

Formula: C₁₃H₂₅NSi
Density: 0.91 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00054932
CAS Number: 87630-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b700d5d2998565ac728148081037669e7145ab85&vid=b700d5d2998565ac728148081037669e7145ab85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR