Tri(2-thienyl)phosphine ≥96.0%

Supplier: TCI America

Synonyms: Tri(thiophen-2-yl)phosphine

T1666-5G T1666-1G
TCT1666-005GEA 851.62 CAD
TCT1666-005G TCT1666-1G
Tri(2-thienyl)phosphine ≥96.0%
Tri(2-thienyl)phosphine

CAS Number: 24171-89-9
MDL Number: MFCD00015471
Molecular Formula: C12H9PS3
Molecular Weight: 280.36
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 205
Freezing point (°C): 25
Storage Temperature: 0-10°C

Formula: C₁₂H₉PS₃
MW: 280.36 g/mol
Boiling Pt: 205 °C
MDL Number: MFCD00015471
CAS Number: 24171-89-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd162b310e726f478d51551f12e3ad911a4011c5&vid=dd162b310e726f478d51551f12e3ad911a4011c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR