Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98.0%

Supplier: TCI America

Synonyms: Tris(2,2'-bipyridyl)dichlororuthenium(II), Dichlorotris(2,2'-bipyridyl)ruthenium(II), Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, Tris(2,2-bipyridyl)ruthenium(II) chloride hexahydrate

T1655-1G T1655-5G
TCT1655-001GEA 207.86 CAD
TCT1655-001G TCT1655-005G
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate ≥98.0%
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate

CAS Number: 50525-27-4
MDL Number: MFCD00149670
Molecular Formula: C30H24Cl2N6Ru
Molecular Weight: 640.53
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: Dichlorotris(2,2'-bipyridyl)ruthenium(II), Tris(2,2'-bipyridyl)dichlororuthenium(II)

Formula: C₃₀H₂₄Cl₂N₆Ru
MDL Number: MFCD00149670
CAS Number: 50525-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609499535d301560608c69e3583fc00c587da4e3&vid=609499535d301560608c69e3583fc00c587da4e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR