Tri(2-furyl)phosphine ≥98.0%

Supplier: TCI America

T1643-5G T1643-1G
TCT1643-005GEA 230.68 CAD
TCT1643-005G TCT1643-001G
Tri(2-furyl)phosphine ≥98.0%
Tri(2-furyl)phosphine

CAS Number: 5518-52-5
MDL Number: MFCD00151857
Molecular Formula: C12H9O3P
Molecular Weight: 232.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 136
Melting point (°C): 63


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands
Other Product Names / Synonyms: TFP

Formula: C₁₂H₉O₃P
Boiling Pt: 136 °C
Melting Pt: 63 °C
MDL Number: MFCD00151857
CAS Number: 5518-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a59699b7d0a1269f1ea8739848a3e0d6422bd900&vid=a59699b7d0a1269f1ea8739848a3e0d6422bd900&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR