Tetraheptylammonium bromide ≥98.0%

Supplier: TCI America

T1602-25G T1602-5G
TCT1602-25GEA 133.83 CAD
TCT1602-25G TCT1602-5G
Tetraheptylammonium bromide ≥98.0%
Tetraheptylammonium bromide

CAS Number: 4368-51-8
MDL Number: MFCD00011861
Molecular Formula: C28H60BrN
Molecular Weight: 490.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 89

Formula: C₂₈H₆₀BrN
MW: 490.7 g/mol
Melting Pt: 96
MDL Number: MFCD00011861
CAS Number: 4368-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364a99e30e6eab3b7340a485aa908d0a7c5f3a3e&vid=364a99e30e6eab3b7340a485aa908d0a7c5f3a3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR