Tri-O-acetyl-D-glucal ≥96.0%
Supplier: TCI America
Synonyms:
(-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate
CAS Number: 2873-29-2
MDL Number: MFCD00063253
Molecular Formula: C12H16O7
Molecular Weight: 272.25
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 55
Flash Point (°C): 110
Specific rotation [a]20/D: -15 deg (C=2, EtOH)
Product Categories: Bioscience, Glucose, Glycals, O-Substituted Sugars
Other Product Names / Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether
Formula:
C₁₂H₁₆O₇ MW: 272.25 g/mol Melting Pt: 55 °C Flash Pt: 110 °C |
MDL Number:
MFCD00063253 CAS Number: 2873-29-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...