1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98.0%

Supplier: TCI America

Synonyms: 4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

T1583-5G T1583-25G
TCT1583-5GEA 56.88 CAD
TCT1583-5G TCT1583-25G
1-(2-Naphthoyl)-3,3,3-trifluoroacetone ≥98.0%
1-(2-Naphthoyl)-3,3,3-trifluoroacetone

CAS Number: 893-33-4
MDL Number: MFCD00054501
Molecular Formula: C14H9F3O2
Molecular Weight: 266.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 74


Product Categories: Synthetic Organic Chemistry, Ligands, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(2-Naphthoyl)-1,1,1-trifluoroacetone

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00054501
CAS Number: 893-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ba81813b3b4c6029e44529de2361140cf4d7c9e&vid=9ba81813b3b4c6029e44529de2361140cf4d7c9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR