(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥95.0%

Supplier: TCI America

Synonyms: (3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

T1515-5G T1515-25G
TCT1515-005GEA 104.28 CAD
TCT1515-005G TCT1515-025G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥95.0%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

CAS Number: 74163-81-8
MDL Number: MFCD00144533
Molecular Formula: C10H11NO2
Molecular Weight: 177.20
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -166 deg (C=1, 1mol/L NaOH sol.)

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
MDL Number: MFCD00144533
CAS Number: 74163-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df87b85376f80a57c99b20bbcd3c20e7921848e4&vid=df87b85376f80a57c99b20bbcd3c20e7921848e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR