Triethylene Glycol Bis(p-toluenesulfonate) ≥97.0%

Supplier: TCI America

Synonyms: Triethylene Glycol Di-p-tosylate, 2,2'-(Ethylenedioxy)diethyl Ditosylate, 1,2-Bis[2-(p-toluenesulfonyloxy)ethoxy]ethane, (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), 1,2-Bis(2-tosyloxyethoxy)ethane, (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(4-methylbenzenesulfonate), 1,2-Bis[2-(p-toluenesulphonyloxy)ethoxy]ethane

T1488-25G T1488-10G
TCT1488-025GEA 206.98 CAD
TCT1488-025G TCT1488-010G
Triethylene Glycol Bis(p-toluenesulfonate) ≥97.0%
Triethylene Glycol Bis(p-toluenesulfonate)

CAS Number: 19249-03-7
MDL Number: MFCD00048096
Molecular Formula: C20H26O8S2
Molecular Weight: 458.54
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 82


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,2-Bis[2-(p-toluenesulfonyloxy)ethoxy]ethane, 1,2-Bis(2-tosyloxyethoxy)ethane, 2,2'-(Ethylenedioxy)diethyl Ditosylate, Triethylene Glycol Di-p-tosylate

Formula: C₂₀H₂₆O₈S₂
MW: 458.54 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00048096
CAS Number: 19249-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4241ddd8c36b2960e5fb0ab962f5e45026e78909&vid=4241ddd8c36b2960e5fb0ab962f5e45026e78909&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR