Tetrabutylammonium tetraphenylborate ≥98.0%

Supplier: TCI America

Synonyms: Tetrabutylammonium Tetraphenylborate

T1480-5G T1480-1G
TCT1480-5GEA 151.68 CAD
TCT1480-5G TCT1480-001G
Tetrabutylammonium tetraphenylborate ≥98.0%
Tetrabutylammonium tetraphenylborate

CAS Number: 15522-59-5
MDL Number: MFCD00011749
Molecular Formula: C40H56BN
Molecular Weight: 561.71
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 235

Formula: C₄₀H₅₆BN
Melting Pt: 235 °C
MDL Number: MFCD00011749
CAS Number: 15522-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d19fd1d287a4578779269ffb0cf06d61c2cc4b55&vid=d19fd1d287a4578779269ffb0cf06d61c2cc4b55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR