Tetrakis(trimethylsilyl)silane ≥98.0%

Supplier: TCI America

T1464-1G T1464-5G
TCT1464-001GEA 48.98 CAD
TCT1464-001G TCT1464-005G
Tetrakis(trimethylsilyl)silane ≥98.0%
Tetrakis(trimethylsilyl)silane

CAS Number: 4098-98-0
MDL Number: MFCD00054859
Molecular Formula: C12H36Si5
Molecular Weight: 320.85
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White


Other Product Names / Synonyms: 1,1,1,3,3,3-Hexamethyl-2,2-bis(trimethylsilyl)trisilane

Formula: C₁₂H₃₆Si₅
MDL Number: MFCD00054859
CAS Number: 4098-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859fc86f48ea09890d0bef8dbfedb2402a2657c5&vid=859fc86f48ea09890d0bef8dbfedb2402a2657c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR