5,6,7,8-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T1403-25G
T1403-5G
TCT1403-25GEA
232.26
CAD
TCT1403-25G
TCT1403-005G
5,6,7,8-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)
5,6,7,8-Tetrahydroquinoxaline
CAS Number: 34413-35-9
MDL Number: MFCD00038078
Molecular Formula: C8H10N2
Molecular Weight: 134.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 85
Flash Point (°C): 90
Specific Gravity (20/20): 1.08
Formula:
C₈H₁₀N₂ Boiling Pt: 85 °C Density: 1.08 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00038078 CAS Number: 34413-35-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...