D-(-)-Diisopropyl tartrate ≥98.0%

Supplier: TCI America

Synonyms: (2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester

T1387-25G T1387-250G
TCT1387-025GEA 102.54 CAD
TCT1387-025G TCT1387-250G
D-(-)-Diisopropyl tartrate ≥98.0%
D(-)-Diisopropyl tartrate

CAS Number: 62961-64-2
MDL Number: MFCD00008876
Molecular Formula: C10H18O6
Molecular Weight: 234.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 110
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: -17 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Tartaric Acid Diisopropyl Ester

Formula: C₁₀H₁₈O₆
MW: 234.25 g/mol
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008876
CAS Number: 62961-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba45e15a30fd461e796daaca6cf5cc6726274e2&vid=3ba45e15a30fd461e796daaca6cf5cc6726274e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR