Benzyltrimethylammonium tribromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T1382-25G T1382-5G
TCT1382-025GEA 88.48 CAD
TCT1382-025G TCT1382-005G
Benzyltrimethylammonium tribromide ≥97.0% (by titrimetric analysis)
Benzyltrimethylammonium tribromide

[Brominating Reagent]
CAS Number: 111865-47-5
MDL Number: MFCD00134423
Molecular Formula: C10H16Br3N
Molecular Weight: 389.96
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 102

Formula: C₁₀H₁₆Br₃N
Melting Pt: 102 °C
MDL Number: MFCD00134423
CAS Number: 111865-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d688bac2ea9376eb310debf2cdc9dfda68926581&vid=88a2356b08a12a6cf30d5b138c552d49dcd9516e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR