S-Furfuryl thioacetate ≥97.0%

Supplier: TCI America

T1283-25G T1283-10G
TCT1283-25GEA 126.4 CAD
TCT1283-25G TCT1283-010G
S-Furfuryl thioacetate ≥97.0%
S-Furfuryl thioacetate

CAS Number: 13678-68-7
MDL Number: MFCD00010083
Molecular Formula: C7H8O2S
Molecular Weight: 156.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 91
Flash Point (°C): 92
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Thioacetic Acid S-Furfuryl Ester

Formula: C₇H₈O₂S
Boiling Pt: 91 °C
Density: 1.16 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00010083
CAS Number: 13678-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=637dd533d131b2169efa5dfe6bc896e09aa600df&vid=637dd533d131b2169efa5dfe6bc896e09aa600df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR