2,4,4,6-Tetrabromo-2,5-cyclohexadienone ≥97.0% (by titrimetric analysis)
Supplier: TCI America
T1235-5G
T1235-25G
TCT1235-5GEA
208.12
CAD
TCT1235-5G
TCT1235-25G
2,4,4,6-Tetrabromo-2,5-cyclohexadienone ≥97.0% (by titrimetric analysis)
2,4,4,6-Tetrabromo-2,5-cyclohexadienone
CAS Number: 20244-61-5
MDL Number: MFCD00001589
Molecular Formula: C6H2Br4O
Molecular Weight: 409.70
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: TBCO
Formula:
C₆H₂Br₄O |
MDL Number:
MFCD00001589 CAS Number: 20244-61-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...