Methyl-2-thiophenecarboxylate ≥99.0%

Supplier: TCI America

Synonyms: Methyl thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Methyl Ester

T1139-5G T1139-25G
TCT1139-5GEA 64.86 CAD
TCT1139-5G TCT1139-25G
Methyl-2-thiophenecarboxylate ≥99.0%
Methyl-2-thiophenecarboxylate

CAS Number: 5380-42-7
MDL Number: MFCD00016895
Molecular Formula: C6H6O2S
Molecular Weight: 142.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 98
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: 2-Thiophenecarboxylic Acid Methyl Ester

Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 98 °C
Density: 1.23 (20 °C)
MDL Number: MFCD00016895
CAS Number: 5380-42-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd9b3df8071ca4767aaead70d73a19ae79fe1e3c&vid=fd9b3df8071ca4767aaead70d73a19ae79fe1e3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR