1,2,3,4-Tetrahydrocarbazole ≥99.0%
Supplier: TCI America
T1006-25G
T1006-100G
TCT1006-025GEA
271.76
CAD
TCT1006-025G
TCT1006-100G
1,2,3,4-Tetrahydrocarbazole ≥99.0%
1,2,3,4-Tetrahydrocarbazole
CAS Number: 942-01-8
MDL Number: MFCD00004959
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 186
Melting point (°C): 119
Formula:
C₁₂H₁₃N Boiling Pt: 186 °C Melting Pt: 119 °C |
MDL Number:
MFCD00004959 CAS Number: 942-01-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...