Tetrathiafulvalene ≥98.0%

Supplier: TCI America

T0980-1G T0980-5G T0980-25G
TCT0980-001GEA 275.88 CAD
TCT0980-001G TCT0980-005G TCT0980-25G
Tetrathiafulvalene ≥98.0%
Tetrathiafulvalene

CAS Number: 31366-25-3
MDL Number: MFCD00005492
Molecular Formula: C6H4S4
Molecular Weight: 204.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 120
Lambda max.: 369 nm (CHCl3)


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: TTF

Formula: C₆H₄S₄
Melting Pt: 120 °C
MDL Number: MFCD00005492
CAS Number: 31366-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=873f56551091dec6425e81c90c521c2dfe179ba4&vid=873f56551091dec6425e81c90c521c2dfe179ba4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR