Tetrahydropyran-2-methanol ≥96.0%

Supplier: TCI America

Synonyms: 2-Hydroxymethyltetrahydropyran, 2-(Hydroxymethyl)tetrahydro-2H-pyran

T0952-25ML T0952-5ML
TCT0952-025MLEA 420.28 CAD
TCT0952-025ML TCT0952-5ML
Tetrahydropyran-2-methanol ≥96.0%
Tetrahydropyran-2-methanol

CAS Number: 100-72-1
MDL Number: MFCD00006624
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 187
Flash Point (°C): 93
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-(Hydroxymethyl)tetrahydropyran, Tetrahydro-2-hydroxymethyl-2H-pyran

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 187 °C
Density: 1.03 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00006624
CAS Number: 100-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70c85dc5895ea8a524c7a2b8cf0dcc7c63f37c7e&vid=70c85dc5895ea8a524c7a2b8cf0dcc7c63f37c7e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR