Trimethylolpropane triacrylate ≥75.0% stabilized

Supplier: TCI America

Synonyms: TMPTA, 1,1,1-Tris(acryloyloxymethyl)propane

T0949-500G T0949-25G
TCT0949-500GEA 102.54 CAD
TCT0949-500G TCT0949-025G
Trimethylolpropane triacrylate ≥75.0% stabilized
Trimethylolpropane triacrylate

(stabilized with MEHQ)
CAS Number: 15625-89-5
MDL Number: MFCD00008628
Molecular Formula: C15H20O6
Molecular Weight: 296.32
Purity/Analysis Method: >75.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 177
Specific Gravity (20/20): 1.11


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: TMPTA, 1,1,1-Tris(acryloyloxymethyl)propane

Formula: C₁₅H₂₀O₆
MW: 296.32 g/mol
Density: 1.11 (20 °C)
Flash Pt: 177 °C
MDL Number: MFCD00008628
CAS Number: 15625-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee1704dee215e1fa5c0fbb57a900d0cd7b31b1b9&vid=ee1704dee215e1fa5c0fbb57a900d0cd7b31b1b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR