Tetraethylammonium toluene-4-sulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0937-25G T0937-500G
TCT0937-025GEA 60.04 CAD
TCT0937-025G TCT0937-500G
Tetraethylammonium toluene-4-sulfonate ≥98.0% (by titrimetric analysis)
Tetraethylammonium toluene-4-sulphonate

CAS Number: 733-44-8
MDL Number: MFCD00011831
Molecular Formula: C15H27NO3S
Molecular Weight: 301.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 114

Formula: C₁₅H₂₇NO₃S
Melting Pt: 114 °C
MDL Number: MFCD00011831
CAS Number: 733-44-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecb7c14256178b709b622897c7098048856eb87d&vid=ecb7c14256178b709b622897c7098048856eb87d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR