Trithiocyanuric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0935-25G T0935-500G T0935-100G
TCT0935-025GEA 75.84 CAD
TCT0935-025G TCT0935-500G TCT0935-100G
Trithiocyanuric acid ≥98.0% (by titrimetric analysis)
Trithiocyanuric acid

CAS Number: 638-16-4
MDL Number: MFCD00006052
Molecular Formula: C3H3N3S3
Molecular Weight: 177.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Slightly Pale Yellow


Other Product Names / Synonyms: 1,3,5-Triazine-2,4,6-trithiol, Trithiocyanuric Acid

Formula: C₃H₃N₃S₃
MDL Number: MFCD00006052
CAS Number: 638-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a30144203f1fb2bdae1fcfadcdc75d2cdb4a53e&vid=8a30144203f1fb2bdae1fcfadcdc75d2cdb4a53e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR