Ethyl thiophene-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-thiophenecarboxylate

T0904-10ML T0904-25ML
TCT0904-010MLEA 34.76 CAD
TCT0904-010ML TCT0904-025ML
Ethyl thiophene-2-carboxylate ≥98.0%
Ethyl thiophene-2-carboxylate

CAS Number: 2810-04-0
MDL Number: MFCD00005436
Molecular Formula: C7H8O2S
Molecular Weight: 156.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 218
Flash Point (°C): 88
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 2-Thiophenecarboxylic Acid Ethyl Ester

Formula: C₇H₈O₂S
MW: 156.20 g/mol
Boiling Pt: 218 °C
Density: 1.16 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00005436
CAS Number: 2810-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff1c4df905ecaf62ad71d4941455351073101ed3&vid=ff1c4df905ecaf62ad71d4941455351073101ed3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR