Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T0806-25G
T0806-5G
TCT0806-025GEA
546.68
CAD
TCT0806-025G
TCT0806-005G
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid ≥98.0% (by titrimetric analysis)
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid
CAS Number: 869-52-3
MDL Number: MFCD00004290
Molecular Formula: C18H30N4O12
Molecular Weight: 494.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: TTHA
Formula:
C₁₈H₃₀N₄O₁₂ |
MDL Number:
MFCD00004290 CAS Number: 869-52-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...