N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T0781-500G T0781-25G
TCT0781-500GEA 127.59 CAD
TCT0781-500G TCT0781-025G
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine ≥98.0% (by titrimetric analysis)
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine

CAS Number: 102-60-3
MDL Number: MFCD00004534
Molecular Formula: C14H32N2O4
Molecular Weight: 292.42
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 211


Other Product Names / Synonyms: Ethylenediamine-N,N,N',N'-tetra-2-propanol, 1,1',1'',1'''-(Ethylenedinitrilo)tetra-2-propanol

Formula: C₁₄H₃₂N₂O₄
Flash Pt: 211 °C
MDL Number: MFCD00004534
CAS Number: 102-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16fd727512b8aaea7538075a020b14f348cfd675&vid=16fd727512b8aaea7538075a020b14f348cfd675&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR