2,3,3-Trimethylindolenine ≥97.0% (by total nitrogen basis)

Supplier: TCI America

T0766-25ML T0766-500ML T0766-100ML
TCT0766-025MLEA 135.88 CAD
TCT0766-025ML TCT0766-500ML TCT0766-100ML
2,3,3-Trimethylindolenine ≥97.0% (by total nitrogen basis)
2,3,3-Trimethylindolenine

CAS Number: 1640-39-7
MDL Number: MFCD00005724
Molecular Formula: C11H13N
Molecular Weight: 159.23
Purity/Analysis Method: >97.0% (N)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 229
Flash Point (°C): 93
Specific Gravity (20/20): 0.99

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 229 °C
Density: 0.99 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00005724
CAS Number: 1640-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87c8a439e22d496d3b85bb9644c3ff46944d7787&vid=87c8a439e22d496d3b85bb9644c3ff46944d7787&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR