Thioisonicotinamide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Pyridine-4-carbothioamide

T0764-25G T0764-5G
TCT0764-25GEA 56.88 CAD
TCT0764-25G TCT0764-005G
Thioisonicotinamide ≥97.0% (by titrimetric analysis)
Thioisonicotinamide

CAS Number: 2196-13-6
MDL Number: MFCD00006437
Molecular Formula: C6H6N2S
Molecular Weight: 138.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 200


Other Product Names / Synonyms: 4-Pyridylthiocarboxamide

Formula: C₆H₆N₂S
MW: 138.19 g/mol
Melting Pt: 200 °C
MDL Number: MFCD00006437
CAS Number: 2196-13-6

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR