Sodium-p-toluenesulfinate tetrahydrate ≥98.0%

Supplier: TCI America

Synonyms: Sodium 4-methylbenzenesulfinate tetrahydrate, p-Toluenesulphinic acid Sodium Salt, Sodium 4-methylbenzenesulphinate tetrahydrate, p-Toluenesulfinic acid Sodium Salt

T0763-500G T0763-25G
TCT0763-500GEA 372.88 CAD
TCT0763-500G TCT0763-025G
Sodium-p-toluenesulfinate tetrahydrate ≥98.0%
Sodium-p-toluenesulfinate tetrahydrate

CAS Number: 868858-48-4
MDL Number: MFCD00149644
Molecular Formula: C7H8O2S
Molecular Weight: 178.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: p-Toluenesulfinic Acid Sodium Salt

Formula: C₇H₁₅NaO₆S
MW: 250.25 g/mol
MDL Number: MFCD00149644
CAS Number: 868858-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f6080357579fb7be8b9165c9170fe2597618ef9&vid=6f6080357579fb7be8b9165c9170fe2597618ef9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR